Learn More
3-Benzyloxy-2-(2-hydroxy-2-phenylethyl)-4H-pyran-4-one, TRC
Product Code. 30330784
Change view
Click to view available options
Kvantitet:
250 mg
Product Code. Kvantitet
30330784 250 mg
1 options
Product Code. 30330784 Supplier Toronto Research Chemicals Supplier No. TRCB287105250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 3-Benzyloxy-2-(2-hydroxy-2-phenylethyl)-4H-pyran-4-one
CAS 1206102-04-6
Molekylformel C20H18O4
Kvantitet 250 mg
Synonym 3-(Benzyloxy)-2-(2-hydroxy-2-phenylethyl)-4-oxo-4H-pyran, 2-(2-Hydroxy-2-phenylethyl)-3-(phenylmethoxy)-4H-pyran-4-one
InChI formel InChI=1S/C20H18O4/c21-17-11-12-23-19(13-18(22)16-9-5-2-6-10-16)20(17)24-14-15-7-3-1-4-8-15/h1-12,18,22H,13-14H2
LEDER O=C1C(OCC2=CC=CC=C2)=C(CC(O)C3=CC=CC=C3)OC=C1
IUPAC-namn 3-(benzyloxy)-2-(2-hydroxy-2-phenylethyl)-4H-pyran-4-one
Molekylvikt (g/mol) 322.35
Formel vikt 322.12