Läs mer
3-Benzyloxy-2-bromo-9H-carbazole N-Carboxylic Acid tert-Butyl Ester, TRC
Produktkod. 30338261
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30338261 100 mg
1 options
Produktkod. 30338261 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB285605100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Benzyloxy-2-bromo-9H-carbazole N-Carboxylic Acid tert-Butyl Ester
CAS 1798043-12-5
Molekylformel C24H22BrNO3
Kvantitet 100 mg
Synonym Tert-butyl 2-bromo-3-phenylmethoxycarbazole-9-carboxylate
InChI formel InChI=1S/C24H22BrNO3/c1-24(2,3)29-23(27)26-20-12-8-7-11-17(20)18-13-22(19(25)14-21(18)26)28-15-16-9-5-4-6-10-16/h4-14H,15H2,1-3H3
LEDER O=C(OC(C)(C)C)N(C1=C2C=C(OCC3=CC=CC=C3)C(Br)=C1)C4=C2C=CC=C4
IUPAC-namn 3-Benzyloxy-2-bromo-9H-carbazole N-Carboxylic Acid tert-Butyl Ester
Molekylvikt (g/mol) 452.34
Formel vikt 451.08