missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybromobenzene, 96%, Thermo Scientific Chemicals

Catalog Number p-4228367
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
53087-13-1
C13H11BrO
263.13
HVWZMGZBJCJDOX-UHFFFAOYSA-N
1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065
2756638
1-bromo-3-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC(=CC=C2)Br
This item is not returnable. View return policy
53087-13-1
C13H11BrO
263.13
HVWZMGZBJCJDOX-UHFFFAOYSA-N
1-benzyloxy-3-bromobenzene, 3-benzyloxybromobenzene, benzyl 3-bromophenyl ether, 1-bromo-3-benzyloxybenzene, 3-benzyloxy bromobenzene, 3-bromophenyl benzyl ether, benzene, 1-bromo-3-phenylmethoxy, 1-benzyloxy-3-bromo-benzene, pubchem3065
2756638
1-bromo-3-phenylmethoxybenzene
C1=CC=C(C=C1)COC2=CC(=CC=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 60°C to 62°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 263.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Benzyloxybromobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Benzyloxybromobenzene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'