missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxypyrrolidine, 96%, Thermo Scientific Chemicals

Catalog Number p-7033625
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
420137-14-0
C11H15NO
177.247
MFCD08060975
CWBMYKUPMLRKQK-UHFFFAOYSA-N
3-benzyloxy pyrrolidine, 3-benzyloxypyrrolidine, 3-benzyloxy-pyrrolidine, 3-phenylmethoxy pyrrolidine, benzyl pyrrolidin-3-yl ether, pyrrolidin-3-yl oxy methyl benzene
14497353
3-phenylmethoxypyrrolidine
C1CNCC1OCC2=CC=CC=C2
This item is not returnable. View return policy
420137-14-0
C11H15NO
177.247
MFCD08060975
CWBMYKUPMLRKQK-UHFFFAOYSA-N
3-benzyloxy pyrrolidine, 3-benzyloxypyrrolidine, 3-benzyloxy-pyrrolidine, 3-phenylmethoxy pyrrolidine, benzyl pyrrolidin-3-yl ether, pyrrolidin-3-yl oxy methyl benzene
14497353
3-phenylmethoxypyrrolidine
C1CNCC1OCC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 177.25
Percent Purity 96%
Chemical Name or Material 3-Benzyloxypyrrolidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'