missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Boc-amino)azetidine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
500 mg
Description
It is an intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate in organic syntheses.
Solubility
Soluble in water. (5g/100ml)
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
It is an intermediate in organic syntheses.
Solubility
Soluble in water. (5g/100ml)
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Specifications
Boiling Point | 268°C |
Solubility Information | Soluble in water. (5g/100ml) |
Formula Weight | 172.23 |
Percent Purity | 98% |
Density | 1.06 |
Quantity | 250 mg |
Flash Point | 116°C (241°F) |
Chemical Name or Material | 3-(Boc-amino)azetidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'