missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Boc-aminomethyl)azetidine hydrochloride, 95%, Thermo Scientific Chemicals
1404.34 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
500 mg
Description
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is a useful building block in the synthesis of polypeptides and other nitrogen containing compounds with potential biological properties.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Solubility Information | Slightly soluble in water. |
IUPAC Name | tert-butyl N-(azetidin-3-ylmethyl)carbamate |
Formula Weight | 222.71 |
Percent Purity | 95% |
Quantity | 250 mg |
Chemical Name or Material | 3-(Boc-aminomethyl)azetidine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'