Läs mer
3-Bodipy-propanoylaminocaproic Acid, N-Hydroxysuccinimide Ester, TRC
Produktkod. 30332112
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30332112 25 mg
1 options
Produktkod. 30332112 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB67493525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Bodipy-propanoylaminocaproic Acid, N-Hydroxysuccinimide Ester
CAS 217190-09-5
Molekylformel C24 H29 B F2 N4 O5
Kvantitet 25 mg
Synonym Boron, [5-[(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)methyl]-N-[6-[(2,5-dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-1H-pyrrole-2-propanamidato-κN1]difluoro-, (T-4)-, (T-4)-[5-[(3,5-Dimethyl-2H-pyrrol-2-ylidene-κN)methyl]-N-[6-[(2,5-dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-1H-pyrrole-2-propanamidato-κN1]difluoroboron, BODIPY FL-X N-succinimidyl ester, BODIPY FL-X,SE, 3-Bodipy-propanoylaminocaproic Acid N-Hydroxysuccinimide Ester
InChI formel InChI=1S/C24H29BF2N4O5/c1-16-14-17(2)29-20(16)15-19-8-7-18(30(19)25(29,26)27)9-10-21(32)28-13-5-3-4-6-24(35)36-31-22(33)11-12-23(31)34/h7-8,14-15H,3-6,9-13H2,1-2H3,(H,28,32)
LEDER Cc1cc(C)n2c1C=C3C=CC(=[N+]3[B-]2(F)F)CCC(=O)NCCCCCC(=O)ON4C(=O)CCC4=O
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 6-[3-(2,2-difluoro-10,12-dimethyl-3-aza-1-azonia-2-boranuidatricyclo[7.3.0.03,7]dodeca-1(12),4,6,8,10-pentaen-4-yl)propanoylamino]hexanoate
Molekylvikt (g/mol) 502.32
Formel vikt 502.2199