missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-2-fluorobenzyl alcohol, 98%

Brand:  Alfa Aesar™ H64371.03

Additional Details : CAS Number : 261723-32-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471539

  • 260.32 SEK / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-2-fluorobenzyl alcohol
205.03
8679241
261723-32-4
MFCD09842441

Specifications

3-Bromo-2-fluorobenzyl alcohol
Solid
104° to 108°C (4mmHg)
205.03
8679241
261723-32-4
98%
1g
MFCD09842441
Videos
SDS