Läs mer
3-Bromo-2-methylpyridine, 97%, Thermo Scientific™
Produktkod. 15473647
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15473647 250 mg 250mg
11362309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15473647 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H27271.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38749-79-0
Molekylformel C6H6BrN
Molekylvikt (g/mol) 172.03
MDL-nummer MFCD00191224
InChI-nyckel AIPWPTPHMIYYOX-UHFFFAOYSA-N
Synonym 3-bromo-2-picoline, 2-methyl-3-bromopyridine, 3-bromo-2-methyl-pyridine, pyridine, 3-bromo-2-methyl, 2-methyl-3-bromo pyridine, pubchem1220, 3-brom-2-methylpyridin, acmc-209j1t, 3-bromo-2-methyl pyridine, ksc497m1l
PubChem CID 1201003
IUPAC-namn 3-bromo-2-methylpyridine
LEDER CC1=C(Br)C=CC=N1
Specifikationer
Kemiskt namn eller material 3-Bromo-2-methylpyridine
Densitet 1.495
Kokpunkt 76°C (17 mmHg)
Flampunkt 79°C (174°F)
Brytningsindex 1.5605
Kvantitet 250 mg
Formel vikt 172.03
Procent renhet 97%