Learn More
3-Bromo-4-hydroxybenzaldehyde, 97+%
Product Code. 11416426
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416426 25 g 25g
11406426 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11416426 Supplier Thermo Scientific Alfa Aesar Supplier No. B24729.14
 Restricted Item
I lager
Add to basket
CAS: 2973-78-6 | C7H5BrO2 | 201.02 g/mol
3-Bromo-4-hydroxybenzaldehyde is an intermediate in organic syntheses, it is used to produce other chemicals like 5-Brom-4-hydroxy-b-nitrostyrol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-4-hydroxybenzaldehyde is an intermediate in organic syntheses, it is used to produce other chemicals like 5-Brom-4-hydroxy-b-nitrostyrol.
Solubility
Soluble in water 1.33 g/L.
Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2973-78-6
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00017348
InChI Key UOTMHAOCAJROQF-UHFFFAOYSA-N
Synonym benzaldehyde, 3-bromo-4-hydroxy, 3-bromo-4-hydroxy-benzaldehyde, 4-hydroxy-3-bromobenzaldehyde, pubchem3778, acmc-209hav, 2-bromo-4-formylphenol, asischem n48923, 3-bromo-4-hydroxybenzaldehye, ksc490m5b, akos akm00117
PubChem CID 76308
IUPAC Name 3-bromo-4-hydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C=C1Br
Specifications
Melting Point 125°C to 128°C
Quantity 25 g
Beilstein 2205318
Sensitivity Air Sensitive
Solubility Information Soluble in water 1.33g/L.
Formula Weight 201.02
Percent Purity ≥97%
Chemical Name or Material 3-Bromo-4-hydroxybenzaldehyde