missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxy-5-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog Number p-7052459
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1373920-70-7
C8H7BrF3NO
270.049
MFCD22201052
ZGHWRZMMLVKQQA-UHFFFAOYSA-N
3-bromo-4-methoxy-5-trifluoromethyl aniline
86277652
3-bromo-4-methoxy-5-(trifluoromethyl)aniline
COC1=C(C=C(C=C1C(F)(F)F)N)Br
This item is not returnable. View return policy
1373920-70-7
C8H7BrF3NO
270.049
MFCD22201052
ZGHWRZMMLVKQQA-UHFFFAOYSA-N
3-bromo-4-methoxy-5-trifluoromethyl aniline
86277652
3-bromo-4-methoxy-5-(trifluoromethyl)aniline
COC1=C(C=C(C=C1C(F)(F)F)N)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2811
Formula Weight 270.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Bromo-4-methoxy-5-(trifluoromethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'