Learn More
Alfa Aesar™ 3-Bromo-5-(2-methoxybenzylcarbamoyl)benzeneboronic acid, 97%
Brand:  Alfa Aesar™ H61308.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15486426
  • 7076.86 SEK / 1g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
3-Bromo-5-(2-methoxybenzylcarbamoyl)benzeneboronic acid
364
MFCD26960668
Specifications
3-Bromo-5-(2-methoxybenzylcarbamoyl)benzeneboronic acid
Solid
364
1g
97%
MFCD26960668