Learn More
3-Bromo-5-(2-pyridyl)-1-phenyl-1,2-dihydropyridin-2-one, TRC
Product Code. 30329955
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
2.5 g
missing translation for 'unitSize'
2.5g
Product Code. Kvantitet unitSize
30329955 2.5 g 2.5g
1 missing translation for 'options'
Product Code. 30329955 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB6872502.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 3-Bromo-5-(2-pyridyl)-1-phenyl-1,2-dihydropyridin-2-one
CAS 381248-06-2
Molekylformel C16H11BrN2O
Kvantitet 2.5 g
Synonym 3-Bromo-1-phenyl-5-(pyridin-2-yl)-2(1H)-pyridone, 5'-Bromo-1'-phenyl-[2,3'-bipyridin]-6'(1'H)-one
InChI formel InChI=1S/C16H11BrN2O/c17-14-10-12(15-8-4-5-9-18-15)11-19(16(14)20)13-6-2-1-3-7-13/h1-11H
LEDER BrC1=CC(=CN(C1=O)c2ccccc2)c3ccccn3
IUPAC-namn 3-bromo-1-phenyl-5-pyridin-2-ylpyridin-2-one
Molekylvikt (g/mol) 327.18
Formel vikt 326.0055