missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzamide, 96%, Thermo Scientific™

Catalog Number 15449388
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
933585-20-7
C7H5BrFNO
218.025
MFCD07780673
CFWMRENFPXKUBG-UHFFFAOYSA-N
43248499
3-bromo-5-fluorobenzamide
C1=C(C=C(C=C1F)Br)C(=O)N
This item is not returnable. View return policy
933585-20-7
C7H5BrFNO
218.025
MFCD07780673
CFWMRENFPXKUBG-UHFFFAOYSA-N
43248499
3-bromo-5-fluorobenzamide
C1=C(C=C(C=C1F)Br)C(=O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 218.02
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromo-5-fluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'