Läs mer
3-Bromo-5-methylbenzeneboronic acid, 97%, Thermo Scientific™
Produktkod. 15479837
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15479837 1 g 1g
15489837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15479837 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H53070.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 849062-36-8
Molekylformel C7H8BBrO2
Molekylvikt (g/mol) 214.85
MDL-nummer MFCD06411355
InChI-nyckel KKEPYOBFWSGWLQ-UHFFFAOYSA-N
Synonym 3-bromo-5-methylphenyl boronic acid, 3-bromo-5-methylbenzeneboronic acid, boronic acid,b-3-bromo-5-methylphenyl, 3-bromo-5-methylphenylboronicacid, acmc-209pyo, 3-bromo-5-methyl-phenyl boronic acid, 3-bromanyl-5-methyl-phenyl boronic acid
PubChem CID 16217686
IUPAC-namn (3-bromo-5-methylphenyl)boronic acid
LEDER CC1=CC(Br)=CC(=C1)B(O)O
Specifikationer
Kemiskt namn eller material 3-Bromo-5-methylbenzeneboronic acid
Smältpunkt 276°C to 278°C
Kvantitet 1 g
Formel vikt 214.85
Procent renhet 97%