missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-nitroanisole, 98%

Brand:  Alfa Aesar™ H64864.06

Additional Details : CAS Number : 16618-67-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15472259

  • 3114.20 SEK / 5g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-nitroanisole
MFCD04112752
1-Bromo-3-methoxy-5-nitrobenzene
16618-67-0
1947332

Specifications

3-Bromo-5-nitroanisole
16618-67-0
Brown
5g
C7H6BrNO3
1947332
MEQKSFQEPDRNEQ-UHFFFAOYSA-N
232.033
232.03
85° to 88°C
Powder
98%
MFCD04112752
1-Bromo-3-methoxy-5-nitrobenzene
COC1=CC(=CC(=C1)[N+](=O)[O-])Br
12299076
Videos
SDS