Läs mer
(+)-3-Bromo-8-camphorsulfonic Acid, TRC
Produktkod. 30335544
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30335544 500 mg
1 options
Produktkod. 30335544 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB682050500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+)-3-Bromo-8-camphorsulfonic Acid
CAS 5344-58-1
Molekylformel C10H15BrO4S
Kvantitet 500 mg
Synonym (+)-alpha-Bromocamphor-π-sulfonic Acid, (+)-alpha-Bromo-camphor-π-sulfonic Acid, R-(+)-3-Bromocamphor-8-sulfonic Acid, d-alpha-Bromocamphor-π-sulfonate, (1S,2S,4R,7R)-2-Bromo-4,7-dimethyl-3-oxo-bicyclo[2.2.1]heptane-7-methanesulfonic Acid
InChI formel InChI=1S/C10H15BrO4S/c1-9-4-3-6(7(11)8(9)12)10(9,2)5-16(13,14)15/h6-7H,3-5H2,1-2H3,(H,13,14,15)
LEDER CC1(CS(=O)(=O)O)[C@@H]2CC[C@@]1(C)C(=O)[C@H]2Br
IUPAC-namn (3-bromo-1,7-dimethyl-2-oxo-7-bicyclo[2.2.1]heptanyl)methanesulfonic acid
Molekylvikt (g/mol) 311.19