missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2,4-dichlorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64925.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449256

  • 2439.30 SEK / 1g


Estimated Shipment date
from Supplier 15-11-2018
Add to basket

Specifications

Chemical Identifiers

MFCD27665325

Specifications

3-Bromo-N-(2,4-dichlorophenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665325
Solid
1g
449.06
97%
Videos
SDS