Learn More
Alfa Aesar™ 3-Bromo-N-(2,5-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64550.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15448976
  • 1966.86 SEK / 1g
Estimated Shipment date
from Supplier 06-03-2018
Add to basket
Specifications
Chemical Identifiers
3-Bromo-N-(2,5-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
416.16
MFCD27665287
Specifications
3-Bromo-N-(2,5-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
1g
416.16
Solid
97%
MFCD27665287