missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-bromo-4-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64015.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448646

  • 1696.90 SEK / 1g


Estimated Shipment date
from Supplier 16-11-2018
Add to basket

Specifications

Chemical Identifiers

MFCD27665243

Specifications

3-Bromo-N-(2-bromo-4-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665243
Solid
1g
477.07
97%
Videos
SDS