Learn More
Alfa Aesar™ 3-Bromo-N-(2-chlorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64344.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15468856
  • 3249.18 SEK / 1g
Estimated Shipment date
from Supplier 06-08-2018
Add to basket
Specifications
Chemical Identifiers
3-Bromo-N-(2-chlorophenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665272
Specifications
3-Bromo-N-(2-chlorophenyl)-5-(trifluoromethyl)benzenesulfonamide
Solid
97%
414.62
1g
MFCD27665272