missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-fluoro-3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64040.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15488656

  • 7780.68 SEK / 5g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2-fluoro-3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
412.2
MFCD27665245

Specifications

3-Bromo-N-(2-fluoro-3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
97%
412.2
Solid
5g
MFCD27665245
Videos
SDS