Learn More
Alfa Aesar™ 3-Bromo-N-[2-fluoro-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64496.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15485685
  • 1937.94 SEK / 1g
Estimated Shipment date
from Supplier 06-08-2018
Add to basket
Specifications
Chemical Identifiers
3-Bromo-N-[2-fluoro-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
MFCD27665285
Specifications
3-Bromo-N-[2-fluoro-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
Solid
1g
466.17
97%
MFCD27665285