Learn More
Alfa Aesar™ 3-Bromo-N-(3,5-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64952.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15469276
  • 7780.68 SEK / 5g
Estimated Shipment date
from Supplier 02-07-2018
Add to basket
Specifications
Chemical Identifiers
3-Bromo-N-(3,5-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
416.16
MFCD27665329
Specifications
3-Bromo-N-(3,5-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
5g
416.16
Solid
97%
MFCD27665329