missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(3,5-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64377.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468876

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(3,5-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide
408.23
MFCD27665275

Specifications

3-Bromo-N-(3,5-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide
1g
408.23
Solid
97%
MFCD27665275
Videos
SDS