Learn More
Alfa Aesar™ 3-Bromo-N-(4-fluoro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64935.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15449266
  • 10070.68 SEK / 5g
Estimated Shipment date
from Supplier 23-01-2019
Add to basket
Specifications
Chemical Identifiers
MFCD27665328
Specifications
3-Bromo-N-(4-fluoro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665328
Solid
5g
412.2
97%