Learn More
Alfa Aesar™ 3-Bromo-N-(4-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64868.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15499206
  • 9009.49 SEK / 5g
Estimated Shipment date
from Supplier 18-02-2019
Add to basket
Specifications
Chemical Identifiers
MFCD27665320
Specifications
3-Bromo-N-(4-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665320
Solid
5g
398.17
97%