Learn More
Alfa Aesar™ 3-Bromo-N-(4-methylbenzyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64582.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15468996
  • 3249.18 SEK / 1g
Estimated Shipment date
from Supplier 04-07-2018
Add to basket
Specifications
Chemical Identifiers
3-Bromo-N-(4-methylbenzyl)-5-(trifluoromethyl)benzenesulfonamide
408.23
MFCD27975682
Specifications
3-Bromo-N-(4-methylbenzyl)-5-(trifluoromethyl)benzenesulfonamide
Solid
408.23
1g
97%
MFCD27975682