missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(5-fluoro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64507.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15498956

  • 9540.08 SEK / 5g


Estimated Shipment date
from Supplier 16-04-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665286

Specifications

3-Bromo-N-(5-fluoro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665286
Solid
5g
412.2
97%
Videos
SDS