missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(5-fluoro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64507.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488956

  • 2159.69 SEK / 1g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(5-fluoro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665286

Specifications

3-Bromo-N-(5-fluoro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
Solid
1g
412.2
97%
MFCD27665286
Videos
SDS