Learn More
3-Bromoacetylpyridine, Hydrobromide, TRC
Product Code. 30334497
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30334497 5 g 5g
1 options
Product Code. 30334497 Supplier Toronto Research Chemicals Supplier No. TRCB6791105G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Bromoacetylpyridine, Hydrobromide
CAS 17694-68-7
Molecular Formula C7 H6 Br N O . Br H
Quantity 5 g
InChI Formula InChI=1S/C7H6BrNO.BrH/c8-4-7(10)6-2-1-3-9-5-6;/h1-3,5H,4H2;1H
SMILES Br.BrCC(=O)c1cccnc1
IUPAC Name 2-bromo-1-pyridin-3-ylethanone;hydrobromide
Molecular Weight (g/mol) 280.94
Formula Weight 278.8894