missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromobenzhydrazide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.
Solubility
Slightly soluble in water (3.7 g/L) (25°C)
Notes
Keep container tightly closed. Store away from oxidizing agents.
3-Bromobenzhydrazide is used to produce 5-(3-bromo-phenyl)-3H-[1,3,4]oxadiazole-2-thione. This reaction will need reagent potassium hydroxide and solvent ethanol.
Solubility
Slightly soluble in water (3.7 g/L) (25°C)
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 154°C to 157°C |
Quantity | 5 g |
Beilstein | 1945814 |
Solubility Information | Slightly soluble in water (3.7g/L) (25°C) |
Formula Weight | 215.05 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-Bromobenzhydrazide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'