Learn More
3-Bromobenzoic acid, 99%, Thermo Scientific Chemicals
Catalog Number p-3756978
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
585-76-2
C7H5BrO2
201.02
MFCD00002487
VOIZNVUXCQLQHS-UHFFFAOYSA-N
m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid
11456
3-bromobenzoic acid
C1=CC(=CC(=C1)Br)C(=O)O
585-76-2
C7H5BrO2
201.02
MFCD00002487
VOIZNVUXCQLQHS-UHFFFAOYSA-N
m-bromobenzoic acid, 3-brombenzoic acid, benzoic acid, 3-bromo, 3-bromo-benzoic acid, benzoic acid, m-bromo, unii-7dct3a73lx, 7dct3a73lx, 3-bromo benzene acid, 3-bromobenzoicacid, 4-fluorobenzoicacid
11456
3-bromobenzoic acid
C1=CC(=CC(=C1)Br)C(=O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 154°C to 158°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula BrC6H4CO2H
Quantity 5 g
Beilstein 09, 349
Solubility Information Solubility in water: 0.54g/L (32°C)
Formula Weight 201.02
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Bromobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.