missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzyl bromide, 99%, Thermo Scientific Chemicals

Catalog Number p-7022350
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
823-78-9
C7H6Br2
249.933
MFCD00000176
ZPCJPJQUVRIILS-UHFFFAOYSA-N
3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
69979
1-bromo-3-(bromomethyl)benzene
C1=CC(=CC(=C1)Br)CBr
This item is not returnable. View return policy
823-78-9
C7H6Br2
249.933
MFCD00000176
ZPCJPJQUVRIILS-UHFFFAOYSA-N
3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
69979
1-bromo-3-(bromomethyl)benzene
C1=CC(=CC(=C1)Br)CBr

3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzyl bromide was used in the synthesis of, 1,7-di(3-bromobenzyl)cyclen and substituted 8-arylquinoline, phosphodiesterase 4 (PDE4) inhibitors.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 40°C to 44°C
Density 1.56
Boiling Point 128°C to 130°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN1759
Beilstein 2078683
Solubility Information Insoluble in water.
Formula Weight 249.94
Percent Purity 99%
Chemical Name or Material 3-Bromobenzyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'