missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-7045870
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
23138-55-8
C7H4BrNO
198.02
MFCD00002014
VQVBCZQTXSHJGF-UHFFFAOYSA-N
3-bromophenyl isocyanate, 3-bromophenylisocyanate, benzene, 1-bromo-3-isocyanato, isocyanic acid 3-bromophenyl ester, 3-bromobenzenisocyanate, m-bromophenylisocyanate, 3-bromophenyliso-cyanate, 3-bromo phenyl isocyanate, 3-bromo-phenyl isocyanate, acmc-209g1f
141001
1-bromo-3-isocyanatobenzene
BrC1=CC=CC(=C1)N=C=O
This item is not returnable. View return policy
23138-55-8
C7H4BrNO
198.02
MFCD00002014
VQVBCZQTXSHJGF-UHFFFAOYSA-N
3-bromophenyl isocyanate, 3-bromophenylisocyanate, benzene, 1-bromo-3-isocyanato, isocyanic acid 3-bromophenyl ester, 3-bromobenzenisocyanate, m-bromophenylisocyanate, 3-bromophenyliso-cyanate, 3-bromo phenyl isocyanate, 3-bromo-phenyl isocyanate, acmc-209g1f
141001
1-bromo-3-isocyanatobenzene
BrC1=CC=CC(=C1)N=C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.586
Boiling Point 220°C
Flash Point 108°C (226°F)
Odor Pungent
Refractive Index 1.5855
Quantity 1 g
UN Number UN2206
Beilstein 2802537
Sensitivity Moisture sensitive
Formula Weight 198.03
Percent Purity 98%
Chemical Name or Material 3-Bromophenyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'