missing translation for 'onlineSavingsMsg'
Learn More

(3-Bromopropoxy)-tert-butyldimethylsilane, 97%, stabilized over sodium carbonate, Thermo Scientific Chemicals

Catalog Number p-4229113
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
89031-84-5, 497-19-8
C9H21BrOSi
253.26
QGMROEZDWJTIDW-UHFFFAOYSA-N
3-bromopropoxy-tert-butyldimethylsilane, 3-bromopropoxy tert-butyl dimethylsilane, 3-bromopropoxy-t-butyldimethylsilane, 3-bromopropoxy-tert-butyl-dimethyl-silane, silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl, acmc-209unq, t-butyl 3-bromopropoxy dimethylsilane, bromopropoxy-tert-butyldimethyl silane
607486
3-bromopropoxy-tert-butyl-dimethylsilane
CC(C)(C)[Si](C)(C)OCCCBr
This item is not returnable. View return policy
89031-84-5, 497-19-8
C9H21BrOSi
253.26
QGMROEZDWJTIDW-UHFFFAOYSA-N
3-bromopropoxy-tert-butyldimethylsilane, 3-bromopropoxy tert-butyl dimethylsilane, 3-bromopropoxy-t-butyldimethylsilane, 3-bromopropoxy-tert-butyl-dimethyl-silane, silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl, acmc-209unq, t-butyl 3-bromopropoxy dimethylsilane, bromopropoxy-tert-butyldimethyl silane
607486
3-bromopropoxy-tert-butyl-dimethylsilane
CC(C)(C)[Si](C)(C)OCCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0930g/mL
Boiling Point 182.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 mL
Specific Gravity 1.093
Formula Weight 253.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (3-Bromopropoxy)-tert-butyldimethylsilane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • (3-Bromopropoxy)-tert-butyldimethylsilane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'