missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyridine, 99%, Thermo Scientific Chemicals

Catalog Number p-76722
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
626-55-1
C5H4BrN
158.00
MFCD00006373
NYPYPOZNGOXYSU-UHFFFAOYSA-N
3-pyridyl bromide, pyridine, 3-bromo, 3-bromo pyridine, 3-bromo-pyridine, 5-bromopyridine, unii-xmn8h2xe9h, 5-bromo pyridine, xmn8h2xe9h, 3-pyridylbromide, 3-bromopryidine
12286
CHEBI:51575
3-bromopyridine
BrC1=CC=CN=C1
This item is not returnable. View return policy
626-55-1
C5H4BrN
158.00
MFCD00006373
NYPYPOZNGOXYSU-UHFFFAOYSA-N
3-pyridyl bromide, pyridine, 3-bromo, 3-bromo pyridine, 3-bromo-pyridine, 5-bromopyridine, unii-xmn8h2xe9h, 5-bromo pyridine, xmn8h2xe9h, 3-pyridylbromide, 3-bromopryidine
12286
CHEBI:51575
3-bromopyridine
BrC1=CC=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Brown or Colorless to Yellow
Density 1.6400g/mL
Boiling Point 173°C
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)Br
Refractive Index 1.5690 to 1.571
Quantity 25 mL
Beilstein 20, 233
Specific Gravity 1.64
Solubility Information Solubility in water: 31g/L (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 158
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Bromopyridine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'