missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotetrahydrofuran, TRC

Product Code. 30348640
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30348640 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348640 Supplier Toronto Research Chemicals Supplier No. TRCB688288100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-Bromotetrahydrofuran
CAS 19311-37-6
Molekylformel C4H7BrO
Kvantitet 100 mg
InChI formel InChI=1S/C4H7BrO/c5-4-1-2-6-3-4/h4H,1-3H2
LEDER BrC1CCOC1
IUPAC-namn 3-bromooxolane
Molekylvikt (g/mol) 151
Formel vikt 149.968
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.