Läs mer
3-Butenyl isothiocyanate, TRC
Produktkod. 30011005
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011005 50 mg
30010924 250 mg
30010523 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011005 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB99910050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3386-97-8
Molekylformel C5H7NS
Molekylvikt (g/mol) 113.18
Synonym Isothiocyanic acid, 3-butenyl ester (6CI,7CI,8CI), 4-Isothiocyanato-1-butene
IUPAC-namn 4-isothiocyanatobut-1-ene
LEDER C=CCCN=C=S
Specifikationer
Kemiskt namn eller material 3-Butenyl isothiocyanate
Kvantitet 50 mg
InChI formel InChI=1S/C5H7NS/c1-2-3-4-6-5-7/h2H,1,3-4H2
Formel vikt 113.0299