missing translation for 'onlineSavingsMsg'
Learn More

3-Carbamoylphenylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 11976031
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
188665-74-9
C13H18BNO3
247.10
MFCD05663886
BPKIPHYWHVOWMS-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-aminocarbonylphenylboronic acid, pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzamide, 3-aminocarbonylphenylboronic acid pinacol ester, benzamide, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 2-dioxaborolan-2-yl-benzamide, bpkiphywhvowms-uhfffaoysa-n, 3-carbamoylphenylboronic acid pinacol ester
2760587
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(N)=O
This item is not returnable. View return policy
188665-74-9
C13H18BNO3
247.10
MFCD05663886
BPKIPHYWHVOWMS-UHFFFAOYSA-N
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 3-aminocarbonylphenylboronic acid, pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzamide, 3-aminocarbonylphenylboronic acid pinacol ester, benzamide, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 2-dioxaborolan-2-yl-benzamide, bpkiphywhvowms-uhfffaoysa-n, 3-carbamoylphenylboronic acid pinacol ester
2760587
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(N)=O
Melting Point 182°C to 183°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 247.1
Percent Purity 97%
Chemical Name or Material 3-Carbamoylphenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-Carbamoylphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'