Läs mer
3-Carboxy Detomidine, TRC
Produktkod. 30136646
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136646 10 mg
30137209 50 mg
30007391 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136646 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17791510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115664-39-6
Molekylformel C12 H12 N2 O2
Molekylvikt (g/mol) 216.24
Synonym 3-(1H-Imidazol-5-ylmethyl)-2-methylbenzoic Acid;
IUPAC-namn 3-(1H-imidazol-4-ylmethyl)-2-methylbenzoic acid
LEDER Cc1c(Cc2c[nH]cn2)cccc1C(=O)O
Specifikationer
Kemiskt namn eller material 3-Carboxy Detomidine
Kvantitet 10 mg
InChI formel InChI=1S/C12H12N2O2/c1-8-9(5-10-6-13-7-14-10)3-2-4-11(8)12(15)16/h2-4,6-7H,5H2,1H3,(H,13,14)(H,15,16)
Formel vikt 216.0899