Läs mer
3-Chloro-10-[3-(dimethylamino)propyl]-9-acridanone, Mikromol™
Produktkod. 30051790
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051790 100 mg
30050976 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051790 Leverantör Mikromol Leverantörsnummer MM0545.13
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Chloro-10-[3-(dimethylamino)propyl]-9-acridanone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Chloro-10-[3-(dimethylamino)propyl]-9-acridanone
CAS 1783028-21-6
Analyt- eller komponentnamn 3-Chloro-10-[3-(dimethylamino)propyl]-9-acridanone
Frakt skick Room Temperature
Molekylformel C18 H19 Cl N2 O
Certifikat och efterlevnad ISO 17025
Synonym 3-Chloro-10-[3-(dimethylamino)propyl]-9-acridanone
InChI formel InChI=1S/C18H19ClN2O/c1-20(2)10-5-11-21-16-7-4-3-6-14(16)18(22)15-9-8-13(19)12-17(15)21/h3-4,6-9,12H,5,10-11H2,1-2H3
LEDER CN(C)CCCN1c2ccccc2C(=O)c3ccc(Cl)cc13
IUPAC-namn 3-chloro-10-[3-(dimethylamino)propyl]acridin-9-one
Molekylvikt (g/mol) 314.81
Kvantitet 100 mg
Formel vikt 314.1186 g/mol