Läs mer
3-Chloro-2,6-dihydroxy-4-methylbenzaldehyde, TRC
Produktkod. 30137548
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137548 50 mg
30133064 10 mg
30011985 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137548 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36416550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57074-21-2
Molekylformel C8 H7 Cl O3
Molekylvikt (g/mol) 186.5924
Synonym 3-Chloro-2,6-dihydroxy-4-methylbenzaldehyde, Chloratranol, Chloroatranol, Benzaldehyde, 3-chloro-2,6-dihydroxy-4-methyl-
IUPAC-namn 3-chloro-2,6-dihydroxy-4-methylbenzaldehyde
LEDER Cc1cc(O)c(C=O)c(O)c1Cl
Specifikationer
Kemiskt namn eller material Chloratranol
Kvantitet 50 mg
InChI formel InChI=1S/C8H7ClO3/c1-4-2-6(11)5(3-10)8(12)7(4)9/h2-3,11-12H,1H3
Formel vikt 186.0084