Läs mer
3-Chloro-2-hydroxypropyltrimethyl Ammonium Chloride (60% in Water), TRC
Produktkod. 30358775
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358775 1 g
30364505 10 g
30358139 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30358775 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3680851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3327-22-8
Molekylformel C6H15Cl2NO
Molekylvikt (g/mol) 188.1
Synonym (2-Hydroxy-3-chloropropyl)trimethylammonium chloride, (3-Chloro-2- hydroxypropyl)-N,N,N-trimethylammonium chloride, 1-Chloro-2-hydroxypropane-3-trimethylammonium chloride, 1-Chloro-3-(trimethylammonio)-2- propanol chloride, 1-Chloro-3-trimethylammonium-2-propanol chloride, COPA 60S, CR 2000, CR 2000 (fixative), Catiomaster C, Catiomaster C 60, Cation Master C, Dextrosil, Dextrosil KA, Dowquat 188, N-(3-Chloro-2-hydroxypropyl)-N,N,N-trimethylammonium chloride, NSC 51216, NT 21, QUAB, QUAB 188, QUAB 218, Quat 188, Reagens CFZ, Reagens S, Reagens S-CFZ, Trimethyl(2-hydroxy-3-chloropropyl)ammonium chloride, Verolan KAF, Vircotone ECO
IUPAC-namn (3-chloro-2-hydroxypropyl)-trimethylazanium;chloride
LEDER [Cl-].C[N+](C)(C)CC(O)CCl
Specifikationer
Kemiskt namn eller material 3-Chloro-2-hydroxypropyltrimethyl Ammonium Chloride
Kvantitet 1 g
InChI formel InChI=1S/C6H15ClNO.ClH/c1-8(2,3)5-6(9)4-7;/h6,9H,4-5H2,1-3H3;1H/q+1;/p-1
Formel vikt 187.0531
Koncentration eller sammansättning (efter analyt eller komponenter) 0.6