Learn More
3-Chloro-2-methyl-1-propene, Sigma-Aldrich™
Product Code. 15859230
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859230 500 mL 500mL
15849230 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15859230 Supplier Sigma Aldrich Supplier No. 8059050500
 Restricted Item
Beräknad leverans: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 563-47-3
Grade Research
Molecular Formula ClCH2C(CH3)=CH2
Molecular Weight (g/mol) 90.55
Specifications
Melting Point -80°C
Density 0.93 g/cm3 at 20°C
MDL Number MFCD00000953
Quantity 500 mL
Synonym 3-Chloro-2-methyl-1-propene, β-Methallyl chloride, 3-Chloroisobutylene, 3-Chloro-2-methyl-1-propene