missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylbenzoic acid, 98%, Thermo Scientific™

Product Code. 11312777
Change view
Click to view available options
Kvantitet:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
11312777 1 g 1g
11322777 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312777 Supplier Thermo Scientific Alfa Aesar Supplier No. L15773.03
Request Bulk or Custom Format

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Detta Thermo Scientific varumärkesprodukt var ursprungligen en del av Alfa Aesar produktportfölj. Viss dokumentation och etikettinformation kan referera till det äldre varumärket. Originalet Alfa Aesar produkt-/artikelkod eller SKU-referens har inte ändrats som en del av varumärkesövergången till Thermo Scientific .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7499-08-3
MDL-nummer MFCD00053312

Specifications

Kemiskt namn eller material 3-Chloro-2-methylbenzoic acid
Smältpunkt 162°C to 166°C
Molekylformel C8H7CIO2
Kvantitet 1 g
Beilstein 1072826
Synonym p-hexadecyloxyaniline, 4-n-hexadecyloxyaniline, 4-hexadecyloxy aniline, 4-hexadecyloxyphenylamine, s-2-4-2,4-diaminopteridin-9-yl methyl methyl amino benzamido pentanedioic acid
InChI-nyckel DETPMDIIFZGDAE-UHFFFAOYSA-N
LEDER CCCCCCCCCCCCCCCCOC1=CC=C(C=C1)N
IUPAC-namn 4-hexadecoxyaniline
Molekylvikt (g/mol) 333.56
PubChem CID 139038
Formel vikt 170.6
Procent renhet 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.