missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-morpholino-1,2,5-thiadiazole, TRC

Product Code. 30364225
Change view
Click to view available options
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30364225 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364225 Supplier Toronto Research Chemicals Supplier No. TRCC379960500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-Chloro-4-morpholino-1,2,5-thiadiazole
CAS 30165-96-9
Molekylformel C6 H8 Cl N3 O S
Kvantitet 500 mg
Synonym 4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine, 3-Chloro-4-morpholino-1,2,5-thiadiazol, Timolol Maleate Imp F (EP)
InChI formel InChI=1S/C6H8ClN3OS/c7-5-6(9-12-8-5)10-1-3-11-4-2-10/h1-4H2
LEDER Clc1nsnc1N2CCOCC2
IUPAC-namn 4-(4-chloro-1,2,5-thiadiazol-3-yl)morpholine
Molekylvikt (g/mol) 205.67
Formel vikt 205.0077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.