Läs mer
3-Chloro-4-nitrophenol, TRC
Produktkod. 30367727
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367727 1 g
30365266 250 mg
30363900 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30367727 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3502751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 491-11-2
Molekylformel C6H4ClNO3
Molekylvikt (g/mol) 173.55
Synonym 2-Chloro-4-hydroxynitrobenzene
IUPAC-namn 3-chloro-4-nitrophenol
LEDER Oc1ccc(c(Cl)c1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 3-Chloro-4-nitrophenol
Kvantitet 1 g
InChI formel InChI=1S/C6H4ClNO3/c7-5-3-4(9)1-2-6(5)8(10)11/h1-3,9H
Formel vikt 172.988