Läs mer
3-chloro-5-fluoro-4-hydroxybenzoic Acid, TRC
Produktkod. 30365299
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30365299 2.5 mg 2.5mg
30353874 5 mg -
30355918 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365299 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3750532.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 455-57-2
Molekylformel C7H4O3FCl
Molekylvikt (g/mol) 190.56
IUPAC-namn 3-chloro-5-fluoro-4-hydroxybenzoic acid
LEDER OC(=O)c1cc(F)c(O)c(Cl)c1
Specifikationer
Kemiskt namn eller material 3-chloro-5-fluoro-4-hydroxybenzoic Acid
Kvantitet 2.5 mg
InChI formel InChI=1S/C7H4ClFO3/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,10H,(H,11,12)
Formel vikt 189.9833