Läs mer
3-Chloro-6-fluoro-2-methoxybenzoic Acid, TRC
Produktkod. 30349039
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349039 100 mg
30353588 500 mg
30365853 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349039 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC585895100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1781848-13-2
Molekylformel C8H6O3FCl
Molekylvikt (g/mol) 204.58
IUPAC-namn 3-chloro-6-fluoro-2-methoxybenzoic acid
LEDER COc1c(Cl)ccc(F)c1C(=O)O
Specifikationer
Kemiskt namn eller material 3-Chloro-6-fluoro-2-methoxybenzoic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C8H6ClFO3/c1-13-7-4(9)2-3-5(10)6(7)8(11)12/h2-3H,1H3,(H,11,12)
Formel vikt 203.9989